3,3,3-Trifluoro-L-lactic acid

Supplier: Apollo Scientific
Danger

Synonyms: (2S)-2-Hydroxy-3,3,3-trifluoropropanoic acid, (2S)-2-Hydroxy-3,3,3-trifluoropropionic acid

PC8122-1G PC8122-250MG
APOSPC8122-1GEA 778 EUR
APOSPC8122-1G APOSPC8122-250MG
3,3,3-Trifluoro-L-lactic acid
3,3,3-Trifluoro-L-lactic acid
Formula: C₃H₃F₃O₃
MW: 144,049 g/mol
Melting Pt: 66…76 °C
Storage Temperature: Ambient
MDL Number: MFCD04037238
CAS Number: 125995-00-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR