4-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid

Supplier: Apollo Scientific

Synonyms: 4-Boronophenyl 4-fluoro-2-nitrophenyl ether

PC8322-1G PC8322-5G
APOSPC8322-1GEA 244 EUR
APOSPC8322-1G APOSPC8322-5G
4-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid
4-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid
Formula: C₁₂H₉BFNO₅
MW: 277,013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800895
CAS Number: 957062-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR