3-Amino-1,1,1-trifluoropropan-2-ol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-3,3,3-trifluoropropylamine, 3-Amino-2-hydroxy-1,1,1-trifluoropropane
PC8329-1G
PC8329-10G
PC8329-5G
PC8329-25G
APOSPC8329-1GEA
52.4
EUR
APOSPC8329-1G
APOSPC8329-10G
APOSPC8329-5G
APOSPC8329-25G
3-Amino-1,1,1-trifluoropropan-2-ol 95%
3-Amino-1,1,1-trifluoropropan-2-ol
Formula:
C₃H₆F₃NO MW: 129,081 g/mol Melting Pt: 99 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD02082679 CAS Number: 431-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...