5-Allyl-2-fluorotoluene
Supplier: Apollo Scientific
Synonyms:
3-(4-Fluoro-3-methylphenyl)prop-1-ene
PC8387-1G
PC8387-5G
APOSPC8387-1GEA
533
EUR
APOSPC8387-1G
APOSPC8387-5G
5-Allyl-2-fluorotoluene
5-Allyl-2-fluorotoluene
Formula:
C₁₀H₁₁F MW: 150,193 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06201193 CAS Number: 842124-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...