2-Fluoro-5-(2-methylallyl)chlorobenzene

Supplier: Apollo Scientific

Synonyms: 3-(3-Chloro-4-fluorophenyl)-2-methylprop-1-ene

PC8398-1G PC8398-5G
APOSPC8398-1GEA 239 EUR
APOSPC8398-1G APOSPC8398-5G
2-Fluoro-5-(2-methylallyl)chlorobenzene
2-Fluoro-5-(2-methylallyl)chlorobenzene
Formula: C₁₀H₁₀ClF
Storage Temperature: Ambient
MDL Number: MFCD07774993
CAS Number: 951888-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR