Pentafluorophenyl pyrimidine-5-carboxylate 97%
Supplier: Apollo Scientific
PC8510-1G
PC8510-250MG
APOSPC8510-1GEA
220
EUR
APOSPC8510-1G
APOSPC8510-250MG
Pentafluorophenyl pyrimidine-5-carboxylate 97%
Pentafluorophenyl pyrimidine-5-carboxylate
Formula:
C₁₁H₃F₅N₂O₂ MW: 290.146 g/mol Melting Pt: 121…123.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD09702344 CAS Number: 921938-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...