4-(Perfluoroethoxy)phenol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Hydroxy-α,α,β,β,β-pentafluorophenetole, 4-(Pentafluoroethoxy)phenol
PC8556-1G
PC8556-250MG
APOSPC8556-1GEA
632
EUR
APOSPC8556-1G
APOSPC8556-250MG
4-(Perfluoroethoxy)phenol
4-(Perfluoroethoxy)phenol
Formula:
C₈H₅F₅O₂ MW: 228,116 g/mol Melting Pt: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00816926 CAS Number: 658-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...