3-Chloro-2,4-difluorobromobenzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-3-chloro-2,4-difluorobenzene
PC8716-1G
PC8716-5G
APOSPC8716-1GEA
21.9
EUR
APOSPC8716-1G
APOSPC8716-5G
3-Chloro-2,4-difluorobromobenzene
3-Chloro-2,4-difluorobromobenzene
Formula:
C₆H₂BrClF₂ Density: 1,8 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142590 CAS Number: 201849-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...