Sodium 2,2,3,3-tetrafluoropropionate 97%
Supplier: Apollo Scientific
|
Danger
|
PC8818-100G
PC8818-25G
APOSPC8818-100GEA
475
EUR
APOSPC8818-100G
APOSPC8818-25G
Sodium 2,2,3,3-tetrafluoropropionate 97%
Sodium 2,2,3,3-tetrafluoropropionate
Formula:
C₃HF₄O₂·Na Storage Temperature: Ambient |
MDL Number:
MFCD00475592 CAS Number: 22898-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...