N,N-Dimethyl-1,1,2,2-tetrafluoroethylamine 95%

Supplier: Apollo Scientific
Danger

PC8821-25G PC8821-5G PC8821-1G
APOSPC8821-25GEA 190 EUR
APOSPC8821-25G APOSPC8821-5G APOSPC8821-1G
N,N-Dimethyl-1,1,2,2-tetrafluoroethylamine 95%
N,N-Dimethyl-1,1,2,2-tetrafluoroethylamine
Formula: C₄H₇F₄N
Boiling Pt: 77 °C (760 mm Hg)
Density: 1,22 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Freezer
MDL Number: MFCD08461915
CAS Number: 1550-50-1
UN: 3129
ADR: 4.3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR