2-Chloroethyl trifluoroacetate 98%

Supplier: Apollo Scientific
Danger

PC8995-25G PC8995-5G
APOSPC8995-25GEA 219 EUR
APOSPC8995-25G APOSPC8995-5G
2-Chloroethyl trifluoroacetate 98%
2-Chloroethyl trifluoroacetate
Formula: C₄H₄ClF₃O₂
Boiling Pt: 121,4 °C
Melting Pt: –116 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00789432
CAS Number: 40949-99-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR