1-Fluoro-2-propoxybenzene 97%
Supplier: Apollo Scientific
PC900046-25G
PC900046-5G
APOSPC900046-25GEA
524
EUR
APOSPC900046-25G
APOSPC900046-5G
1-Fluoro-2-propoxybenzene 97%
1-Fluoro-2-propoxybenzene
Formula:
C₉H₁₁FO MW: 154,181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01861233 CAS Number: 203115-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...