3-(2-fluorophenyl)propan-1-ol 96%
Supplier: Apollo Scientific
PC900075-1G
PC900075-250MG
APOSPC900075-1GEA
632
EUR
APOSPC900075-1G
APOSPC900075-250MG
3-(2-fluorophenyl)propan-1-ol 96%
3-(2-fluorophenyl)propan-1-ol
Formula:
C₉H₁₁FO MW: 154,181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09028696 CAS Number: 76727-24-7 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...