[2-Amino-1-(4-fluorophenyl)ethyl]dimethylamine 96%
Supplier: Apollo Scientific
PC900188-1G
PC900188-250MG
APOSPC900188-1GEA
364
EUR
APOSPC900188-1G
APOSPC900188-250MG
[2-Amino-1-(4-fluorophenyl)ethyl]dimethylamine 96%
[2-Amino-1-(4-fluorophenyl)ethyl]dimethylamine
Formula:
C₁₀H₁₅FN₂ Storage Temperature: Ambient |
MDL Number:
MFCD06753109 CAS Number: 842954-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...