2-(2-Fluorophenyl)-N-methylacetamide 96%
Supplier: Apollo Scientific
PC900231-1G
PC900231-5G
APOSPC900231-1GEA
270
EUR
APOSPC900231-1G
APOSPC900231-5G
2-(2-Fluorophenyl)-N-methylacetamide 96%
2-(2-Fluorophenyl)-N-methylacetamide
Formula:
C₉H₁₀FNO MW: 167,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19595581 CAS Number: 612487-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...