1-(5-Fluoropyridin-2-yl)ethanone 98%
Supplier: Apollo Scientific
PC900560-1G
PC900560-250MG
APOSPC900560-1GEA
328
EUR
APOSPC900560-1G
APOSPC900560-250MG
1-(5-Fluoropyridin-2-yl)ethanone 98%
1-(5-Fluoropyridin-2-yl)ethanone
Formula:
C₇H₆FNO MW: 139,127 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10698609 CAS Number: 915720-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...