5-Chloro-3-fluoro-2-(2-hydroxyethylamino)pyridine 97%
Supplier: Apollo Scientific
PC900569-1G
PC900569-25G
PC900569-5G
APOSPC900569-1GEA
125
EUR
APOSPC900569-1G
APOSPC900569-25G
APOSPC900569-5G
5-Chloro-3-fluoro-2-(2-hydroxyethylamino)pyridine 97%
5-Chloro-3-fluoro-2-(2-hydroxyethylamino)pyridine
Formula:
C₇H₈ClFN₂O MW: 190,603 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18434475 CAS Number: 1280786-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...