2-(5-Chloro-3-fluoropyridin-2-yl)acetic acid 98%

Supplier: Apollo Scientific

PC900577-1G PC900577-250MG
APOSPC900577-1GEA 502 EUR
APOSPC900577-1G APOSPC900577-250MG
2-(5-Chloro-3-fluoropyridin-2-yl)acetic acid 98%
2-(5-Chloro-3-fluoropyridin-2-yl)acetic acid
Formula: C₇H₅ClFNO₂
MW: 189,572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194517
CAS Number: 1214323-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR