2-Ethoxy-5-fluoropyridine 97%

Supplier: Apollo Scientific

PC900585-1G PC900585-5G
APOSPC900585-1GEA 87 EUR
APOSPC900585-1G APOSPC900585-5G
2-Ethoxy-5-fluoropyridine 97%
2-Ethoxy-5-fluoropyridine
Formula: C₇H₈FNO
MW: 141,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18914532
CAS Number: 858675-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR