6-Ethoxy-2-fluoro-3-methylphenylboronic acid 98%

Supplier: Apollo Scientific

PC900897-1G PC900897-25G PC900897-5G
APOSPC900897-1GEA 145 EUR
APOSPC900897-1G APOSPC900897-25G APOSPC900897-5G
6-Ethoxy-2-fluoro-3-methylphenylboronic acid 98%
6-Ethoxy-2-fluoro-3-methylphenylboronic acid
Formula: C₉H₁₂BFO₃
MW: 197,999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981512
CAS Number: 1451391-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR