5-Cyclopropyl-2-fluorophenylboronic acid 95%

Supplier: Apollo Scientific

PC900964-1G PC900964-250MG
APOSPC900964-1GEA 538 EUR
APOSPC900964-1G APOSPC900964-250MG
5-Cyclopropyl-2-fluorophenylboronic acid 95%
5-Cyclopropyl-2-fluorophenylboronic acid
Formula: C₉H₁₀BFO₂
MW: 179.984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21099634
CAS Number: 1586045-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR