1-(4-Fluoro-phenyl)-1H-pyrrole-2-carbaldehyde 95%

Supplier: Apollo Scientific

Synonyms: 1-(4-Fluorophenyl)-1H-pyrrole-2-carbaldehyde

PC901133-1G PC901133-250MG
APOSPC901133-1GEA 546 EUR
APOSPC901133-1G APOSPC901133-250MG
1-(4-Fluoro-phenyl)-1H-pyrrole-2-carbaldehyde 95%
1-(4-Fluoro-phenyl)-1H-pyrrole-2-carbaldehyde
Formula: C₁₁H₈FNO
MW: 189,186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664892
CAS Number: 169036-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR