(3-Chloro-2-fluoro-5-methylphenyl)boronic acid 98%
Supplier: Apollo Scientific
PC901210-1G
PC901210-5G
APOSPC901210-1GEA
69.6
EUR
APOSPC901210-1G
APOSPC901210-5G
(3-Chloro-2-fluoro-5-methylphenyl)boronic acid 98%
(3-Chloro-2-fluoro-5-methylphenyl)boronic acid
Formula:
C₇H₇BClFO₂ MW: 188,392 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22571810 CAS Number: 352535-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...