2-Fluoro-6-propoxypyridine-3-boronic acid 98%

Supplier: Apollo Scientific

Synonyms: (2-Fluoro-6-propoxypyridin-3-yl)boronic acid

PC901347-1G PC901347-5G
APOSPC901347-1GEA 128 EUR
APOSPC901347-1G APOSPC901347-5G
2-Fluoro-6-propoxypyridine-3-boronic acid 98%
2-Fluoro-6-propoxypyridine-3-boronic acid
Formula: C₈H₁₁BFNO₃
MW: 198,987 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18434411
CAS Number: 2096338-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR