5-Fluoro-2,3-dihydroisoindol-1-one 95%

Supplier: Apollo Scientific

Synonyms: 5-Fluoroisoindolin-1-one

PC901570-1G PC901570-250MG
APOSPC901570-1GEA 250 EUR
APOSPC901570-1G APOSPC901570-250MG
5-Fluoro-2,3-dihydroisoindol-1-one 95%
5-Fluoro-2,3-dihydroisoindol-1-one
Formula: C₈H₆FNO
MW: 151,138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18204658
CAS Number: 1260666-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR