2-Fluoro-3-methoxypyridine-5-boronic acid 95%

Supplier: Apollo Scientific

PC901701-1G PC901701-250MG
APOSPC901701-1GEA 379 EUR
APOSPC901701-1G APOSPC901701-250MG
2-Fluoro-3-methoxypyridine-5-boronic acid 95%
2-Fluoro-3-methoxypyridine-5-boronic acid
Formula: C₆H₇BFNO₃
MW: 170,934 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17012001
CAS Number: 1451392-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR