3-Fluoro-2-(pyrrolidin-1-yl)pyridine 98%

Supplier: Apollo Scientific

PC901823-1G PC901823-5G
APOSPC901823-1GEA 168 EUR
APOSPC901823-1G APOSPC901823-5G
3-Fluoro-2-(pyrrolidin-1-yl)pyridine 98%
3-Fluoro-2-(pyrrolidin-1-yl)pyridine
Formula: C₉H₁₁FN₂
MW: 166,195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855858
CAS Number: 1133115-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR