2-(2-Fluorophenyl)propan-2-ol 98%
Supplier: Apollo Scientific
PC902056-1G
PC902056-5G
APOSPC902056-1GEA
168
EUR
APOSPC902056-1G
APOSPC902056-5G
2-(2-Fluorophenyl)propan-2-ol 98%
2-(2-Fluorophenyl)propan-2-ol
Formula:
C₉H₁₁FO MW: 154,181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11870101 CAS Number: 320-12-7 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...