2-(3-Fluorophenyl)propan-2-ol 97%
Supplier: Apollo Scientific
PC902057-1G
PC902057-25G
PC902057-5G
APOSPC902057-1GEA
147
EUR
APOSPC902057-1G
APOSPC902057-25G
APOSPC902057-5G
2-(3-Fluorophenyl)propan-2-ol 97%
2-(3-Fluorophenyl)propan-2-ol
Formula:
C₉H₁₁FO MW: 154,181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11870100 CAS Number: 401-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...