1-Ethyl-3-(trifluoromethyl)pyrazole 98%

Supplier: Apollo Scientific

PC902059-1G PC902059-5G
APOSPC902059-1GEA 219 EUR
APOSPC902059-1G APOSPC902059-5G
1-Ethyl-3-(trifluoromethyl)pyrazole 98%
1-Ethyl-3-(trifluoromethyl)pyrazole
Formula: C₆H₇F₃N₂
MW: 164,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15976388
CAS Number: 362640-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR