5-Amino-3-(3-fluorophenyl)-1-methylpyrazole 95%

Supplier: Apollo Scientific

PC902064-1G PC902064-5G
APOSPC902064-1GEA 219 EUR
APOSPC902064-1G APOSPC902064-5G
5-Amino-3-(3-fluorophenyl)-1-methylpyrazole 95%
5-Amino-3-(3-fluorophenyl)-1-methylpyrazole
Formula: C₁₀H₁₀FN₃
MW: 191,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738294
CAS Number: 1017781-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR