1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Supplier: Apollo Scientific
|
Warning
|
PC902100-1G
PC902100-25G
PC902100-5G
APOSPC902100-1GEA
197
EUR
APOSPC902100-1G
APOSPC902100-25G
APOSPC902100-5G
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formula:
C₆H₅F₃N₂O MW: 178,112 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09991529 CAS Number: 128225-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...