2-(3-Fluorophenyl)benzonitrile 98%

Supplier: Apollo Scientific

PC902361-1G PC902361-5G
APOSPC902361-1GEA 145 EUR
APOSPC902361-1G APOSPC902361-5G
2-(3-Fluorophenyl)benzonitrile 98%
2-(3-Fluorophenyl)benzonitrile
Formula: C₁₃H₈FN
MW: 197,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05980284
CAS Number: 1352318-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR