4-(3-Fluorophenyl)benzonitrile 95%

Supplier: Apollo Scientific

PC902384-1G PC902384-5G
APOSPC902384-1GEA 145 EUR
APOSPC902384-1G APOSPC902384-5G
4-(3-Fluorophenyl)benzonitrile 95%
4-(3-Fluorophenyl)benzonitrile
Formula: C₁₃H₈FN
MW: 197,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05981077
CAS Number: 893734-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR