3-(3-Fluorophenyl)benzonitrile 98%
Supplier: Apollo Scientific
PC902445-1G
PC902445-5G
APOSPC902445-1GEA
219
EUR
APOSPC902445-1G
APOSPC902445-5G
3-(3-Fluorophenyl)benzonitrile 98%
3-(3-Fluorophenyl)benzonitrile
Formula:
C₁₃H₈FN MW: 197,208 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05981494 CAS Number: 893734-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...