1-(3-Fluorophenyl)pyrazole 97%

Supplier: Apollo Scientific

PC902595-1G PC902595-5G
APOSPC902595-1GEA 203 EUR
APOSPC902595-1G APOSPC902595-5G
1-(3-Fluorophenyl)pyrazole 97%
1-(3-Fluorophenyl)pyrazole
Formula: C₉H₇FN₂
MW: 162,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699188
CAS Number: 37649-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR