Ethyl 2-(4-amino-2-fluorophenyl)acetate 97%

Supplier: Apollo Scientific

PC902634-1G PC902634-5G
APOSPC902634-1GEA 186 EUR
APOSPC902634-1G APOSPC902634-5G
Ethyl 2-(4-amino-2-fluorophenyl)acetate 97%
Ethyl 2-(4-amino-2-fluorophenyl)acetate
Formula: C₁₀H₁₂FNO₂
MW: 197,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20484495
CAS Number: 73781-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR