2-Amino-4,6-difluoropyrimidine 95%

Supplier: Apollo Scientific
Warning

Synonyms: 4,6-Difluoropyrimidin-2-amine

PC902684-1G PC902684-5G
APOSPC902684-1GEA 105 EUR
APOSPC902684-1G APOSPC902684-5G
2-Amino-4,6-difluoropyrimidine 95%
2-Amino-4,6-difluoropyrimidine
Formula: C₄H₃F₂N₃
MW: 131.084 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00233527
CAS Number: 675-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR