4-Amino-2,6-difluoropyrimidine 95%
Supplier: Apollo Scientific
PC902691-1G
PC902691-250MG
APOSPC902691-1GEA
115
EUR
APOSPC902691-1G
APOSPC902691-250MG
4-Amino-2,6-difluoropyrimidine 95%
4-Amino-2,6-difluoropyrimidine
Formula:
C₄H₃F₂N₃ Storage Temperature: Ambient |
MDL Number:
MFCD00457061 CAS Number: 675-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...