1-(2-Fluoroethoxy)-2-nitrobenzene 98%
Supplier: Apollo Scientific
PC902706-1G
PC902706-25G
PC902706-5G
APOSPC902706-1GEA
145
EUR
APOSPC902706-1G
APOSPC902706-25G
APOSPC902706-5G
1-(2-Fluoroethoxy)-2-nitrobenzene 98%
1-(2-Fluoroethoxy)-2-nitrobenzene
Formula:
C₈H₈FNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22375007 CAS Number: 132838-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...