5-Fluoro-4-methyl-2-nitrophenol 98%
Supplier: Apollo Scientific
PC902869-1G
PC902869-250MG
APOSPC902869-1GEA
364
EUR
APOSPC902869-1G
APOSPC902869-250MG
5-Fluoro-4-methyl-2-nitrophenol 98%
5-Fluoro-4-methyl-2-nitrophenol
Formula:
C₇H₆FNO₃ MW: 171,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16658619 CAS Number: 83341-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...