6-Chloro-5-fluorobenzotriazole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chloro-5-fluoro-1H-benzo[d][1,2,3]triazole

PC9029-1G PC9029-5G PC9029-10G
APOSPC9029-1GEA 132 EUR
APOSPC9029-1G APOSPC9029-5G APOSPC9029-10G
6-Chloro-5-fluorobenzotriazole 97%
6-Chloro-5-fluorobenzotriazole
Formula: C₆H₃ClFN₃
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00173671
CAS Number: 99803-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR