1-(Allyloxymethyl)-2-fluorobenzene 98%
Supplier: Apollo Scientific
PC902975-1G
PC902975-5G
APOSPC902975-1GEA
147
EUR
APOSPC902975-1G
APOSPC902975-5G
1-(Allyloxymethyl)-2-fluorobenzene 98%
1-(Allyloxymethyl)-2-fluorobenzene
Formula:
C₁₀H₁₁FO MW: 166,192 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13195729 CAS Number: 1199773-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...