5-Fluoro-1H-indazole-3-carbaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC903633-100MG
PC903633-50MG
APOSPC903633-100MGEA
46.4
EUR
APOSPC903633-100MG
APOSPC903633-50MG
5-Fluoro-1H-indazole-3-carbaldehyde 98%
5-Fluoro-1H-indazole-3-carbaldehyde
Formula:
C₈H₅FN₂O MW: 164,137 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06739142 CAS Number: 485841-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...