4-Chloro-6-fluoro-2-methylquinazoline 98%

Supplier: Apollo Scientific

PC903669-1G PC903669-5G
APOSPC903669-1GEA 98.6 EUR
APOSPC903669-1G APOSPC903669-5G
4-Chloro-6-fluoro-2-methylquinazoline 98%
4-Chloro-6-fluoro-2-methylquinazoline
Formula: C₉H₆ClFN₂
MW: 196,609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11047031
CAS Number: 1044768-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR