4-Chloro-2-fluoronicotinaldehyde 95+%
Supplier: Apollo Scientific
|
Warning
|
PC903671-1G
PC903671-250MG
PC903671-5G
APOSPC903671-1GEA
102
EUR
APOSPC903671-1G
APOSPC903671-250MG
APOSPC903671-5G
4-Chloro-2-fluoronicotinaldehyde 95+%
4-Chloro-2-fluoronicotinaldehyde
Formula:
C₆H₃ClFNO Storage Temperature: Refrigerator |
MDL Number:
MFCD15143389 CAS Number: 1232432-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...