N-(5-Fluoro-2-methyl-phenyl)-formamide 95%

Supplier: Apollo Scientific

PC903761-1G PC903761-5G
APOSPC903761-1GEA 219 EUR
APOSPC903761-1G APOSPC903761-5G
N-(5-Fluoro-2-methyl-phenyl)-formamide 95%
N-(5-Fluoro-2-methyl-phenyl)-formamide
Formula: C₈H₈FNO
MW: 153,154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19441849
CAS Number: 366-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR