5-Fluoro-2,3-dihydro-1H-inden-1-ol 95%

Supplier: Apollo Scientific

PC903825-1G PC903825-250MG
APOSPC903825-1GEA 150 EUR
APOSPC903825-1G APOSPC903825-250MG
5-Fluoro-2,3-dihydro-1H-inden-1-ol 95%
5-Fluoro-2,3-dihydro-1H-inden-1-ol
Formula: C₉H₉FO
MW: 152,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11219501
CAS Number: 52085-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR