Potassium (benzyloxycarbonylamino)methyltrifluoroborate

Supplier: Apollo Scientific
Warning

PC904164-1G PC904164-5G
APOSPC904164-1GEA 314 EUR
APOSPC904164-1G APOSPC904164-5G
Potassium (benzyloxycarbonylamino)methyltrifluoroborate
Potassium (benzyloxycarbonylamino)methyltrifluoroborate
Formula: C₉H₁₀BF₃NO₂·K
MW: 271,086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19982304
CAS Number: 1695529-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR