5-Fluoro-N-methoxy-N-methylnicotinamide 95%

Supplier: Apollo Scientific

PC904193-1G PC904193-250MG
APOSPC904193-1GEA 364 EUR
APOSPC904193-1G APOSPC904193-250MG
5-Fluoro-N-methoxy-N-methylnicotinamide 95%
5-Fluoro-N-methoxy-N-methylnicotinamide
Formula: C₈H₉FN₂O₂
MW: 184,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256732
CAS Number: 342602-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR